Assay Details
Target Gene Details
Entrez Gene ID: | 68545 |
Gene Name: | endothelial cell surface expressed chemotaxis and apoptosis regulator |
Gene Aliases: |
1110006O17Rik, ARIA |
Location: |
Chr.18:35713086-35722725 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ecscr | NM_001033141.1 | 9 | 945 | NP_001028313.1 |
| AK003507.1 | 5 | 438 | ||
| AK157485.1 | 9 | 945 | BAE34097.1 |

Genomic Map