Assay Details
Target Gene Details
Entrez Gene ID: | 13653 |
Gene Name: | early growth response 1 |
Gene Aliases: |
A530045N19Rik, ETR103, Egr-1, Krox-1, Krox-24, Krox24, NGF1-A, NGFI-A, NGFIA, TIS8, Zenk, Zfp-6, Zif268, egr |
Location: |
Chr.18:34861207-34864956 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Egr1 | NM_007913.5 | 2 | 1827 | NP_031939.1 |
| AK037837.1 | 3 | 1804 | BAC29885.1 | |
| AK040925.1 | 2 | 1827 | BAC30748.1 | |
| AK133630.1 | 2 | 1325 | BAE21758.1 | |
| AK145198.1 | 2 | 1826 | BAE26289.1 | |
| AK146322.1 | 2 | 1827 | BAE27077.1 | |
| BC138613.1 | 2 | 1616 | ||
| BC138615.1 | 2 | 1616 | ||
| M19643.1 | 2 | 1399 | AAA39382.1 | |
| M20157.1 | 2 | 1803 | AAA37544.1 |
More Information
Set Membership: |
|

Genomic Map