Assay Details
Target Gene Details
Entrez Gene ID: | 13163 |
Gene Name: | Fas death domain-associated protein |
Gene Aliases: |
- |
Location: |
Chr.17:33909381-33915590 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Daxx | NM_001199733.1 | 6 | 1838 | NP_001186662.1 |
| NM_007829.4 | 6 | 1738 | NP_031855.3 | |
| AF006040.1 | 5 | 1589 | AAC53284.1 | |
| AK090356.1 | 7 | 1546 | ||
| AK133863.1 | 6 | 1646 | BAE21895.1 | |
| AK144332.1 | 6 | 1671 | ||
| AK145906.1 | 6 | 1669 | BAE26741.1 | |
| AK146743.1 | 6 | 1665 | BAE27403.1 | |
| AK151279.1 | 6 | 1659 | ||
| AK153149.1 | 6 | 1626 | BAE31759.1 | |
| AK156425.1 | 6 | 1836 | BAE33708.1 | |
| AK172616.1 | 6 | 1839 | ||
| AK213512.1 | 2 | 165 | ||
| AY734459.1 | 4 | 1302 | AAU87500.1 | |
| AY734463.1 | AAU50682.1 | |||
| AY734465.1 | 4 | 1213 | AAU45095.1 | |
| AY758274.1 | AAU84993.1 | |||
| BC128373.1 | 6 | 1648 | ||
| KJ650207.1 | 5 | 1565 |

Genomic Map