Assay Details
Target Gene Details
Entrez Gene ID: | 15002 |
Gene Name: | histocompatibility 2, O region beta locus |
Gene Aliases: |
A-beta-2, A-beta2, H-2Ab, H-2I, H-2Ob, H2-Ab, H2-Ab2, H2-IAb2, Ob |
Location: |
Chr.17:34238895-34245908 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2-Ob | NM_010389.3 | 3 | 567 | NP_034519.2 |
| AK053233.1 | 3 | 616 | ||
| AK155144.1 | 3 | 517 | BAE33074.1 | |
| AK155650.1 | 3 | 567 | BAE33366.1 | |
| AK172191.1 | 3 | 572 | ||
| AK172273.1 | 3 | 464 | BAE42918.1 | |
| AK172299.1 | 3 | 464 | BAE42933.1 | |
| BC100707.1 | 3 | 496 | AAI00708.1 | |
| BC100709.1 | 3 | 493 | AAI00710.1 | |
| BC100710.1 | 3 | 495 | AAI00711.1 | |
| BC127027.1 | 3 | 500 | ||
| BC127028.1 | 3 | 500 | ||
| KJ650230.1 | 3 | 500 | ||
| KJ663722.1 | 3 | 422 |
More Information
Set Membership: |
|

Genomic Map