Assay Details
Target Gene Details
Entrez Gene ID: | 15002 |
Gene Name: | histocompatibility 2, O region beta locus |
Gene Aliases: |
A-beta-2, A-beta2, H-2Ab, H-2I, H-2Ob, H2-Ab, H2-Ab2, H2-IAb2, Ob |
Location: |
Chr.17:34238895-34245908 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2-Ob | NM_010389.3 | 6 | 1145 | NP_034519.2 |
| AK053233.1 | 6 | 1243 | ||
| AK155144.1 | 6 | 1095 | BAE33074.1 | |
| AK155650.1 | 6 | 1145 | BAE33366.1 | |
| AK172191.1 | 6 | 1151 | ||
| AK172273.1 | BAE42918.1 | |||
| AK172299.1 | BAE42933.1 |
More Information
Set Membership: |
|

Genomic Map