Assay Details
Target Gene Details
Entrez Gene ID: | 15006 |
Gene Name: | histocompatibility 2, Q region locus 1 |
Gene Aliases: |
H-2Q1, Q1, Q1b, Q1d, Q1k, Qa-1, Qa1, Qed-1 |
Location: |
Chr.17:35320516-35327699 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2-Q1 | NM_010390.3 | 3 | 522 | NP_034520.2 |
| BC080812.1 | 3 | 533 | AAH80812.1 | |
| U96752.1 | 3 | 522 | AAB71648.1 |
More Information
Set Membership: |
|

Genomic Map