Assay Details
Target Gene Details
Entrez Gene ID: | 80978 |
Gene Name: | MAS-related GPR, member H |
Gene Aliases: |
Gpr90, Mgrb, MrgH |
Location: |
Chr.17:12876034-12877842 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrgprh | NM_030726.1 | 1 | 1205 | NP_109651.1 |
| AF295365.1 | 1 | 1205 | AAK01865.1 | |
| BC116696.1 | 1 | 835 | ||
| BC117525.1 | 1 | 833 |
More Information
Set Membership: |
|

Genomic Map