Assay Details
Target Gene Details
Entrez Gene ID: | 73836 |
Gene Name: | solute carrier family 35, member B2 |
Gene Aliases: |
1110003M08Rik, AA407792, AI842275, PAPST1, Slc35b1 |
Location: |
Chr.17:45563879-45567671 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc35b2 | NM_028662.2 | 1 | 416 | NP_082938.1 |
| AF414190.1 | AAL07487.1 | |||
| AK003377.1 | BAB22749.1 | |||
| AK090329.1 | 1 | 604 | BAC41173.1 | |
| AK144403.1 | BAE25873.1 | |||
| AK152362.1 | BAE31152.1 | |||
| AK154868.1 | BAE32889.1 | |||
| AK155551.1 | BAE33321.1 | |||
| AK167343.1 | 1 | 417 | BAE39443.1 | |
| BC025875.1 | AAH25875.1 | |||
| BC036992.1 | AAH36992.1 |
More Information
Set Membership: |
|

Genomic Map