Assay Details
Target Gene Details
Entrez Gene ID: | 100169874 |
Gene Name: | predicted gene 11110 |
Gene Aliases: |
ENSMUSG00000079414 |
Location: |
Chr.17:57092023-57105942 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm11110 | NR_033508.1 | |||
| AK154177.1 | BAE32422.1 |
Target Gene Details
Entrez Gene ID: | 21950 |
Gene Name: | tumor necrosis factor (ligand) superfamily, member 9 |
Gene Aliases: |
4-1BB-L, 4-1BBL, AI848817, Cd137l, Ly63l |
Location: |
Chr.17:57105277-57107757 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfsf9 | NM_009404.3 | 1 | 234 | NP_033430.1 |
| AK155610.1 | 1 | 235 | BAE33344.1 | |
| AK169471.1 | 1 | 207 | BAE41192.1 | |
| BC138767.1 | 1 | 213 | ||
| BC138770.1 | 1 | 213 |
More Information
Set Membership: |
|

Genomic Map