Assay Details
Target Gene Details
Entrez Gene ID: | 21950 |
Gene Name: | tumor necrosis factor (ligand) superfamily, member 9 |
Gene Aliases: |
4-1BB-L, 4-1BBL, AI848817, Cd137l, Ly63l |
Location: |
Chr.17:57105277-57107757 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfsf9 | NM_009404.3 | 3 | 754 | NP_033430.1 |
| AK155610.1 | 3 | 755 | BAE33344.1 | |
| AK169471.1 | 3 | 727 | BAE41192.1 | |
| BC138767.1 | 3 | 733 | ||
| BC138770.1 | 3 | 733 |

Genomic Map