Assay Details
Target Gene Details
Entrez Gene ID: | 13824 |
Gene Name: | erythrocyte protein band 4.1 like 4a |
Gene Aliases: |
Epb4.1l4, Epb41l4a, NBL4 |
Location: |
Chr.18:33796327-34007678 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epb4.1l4a | NM_013512.2 | 2 | 656 | NP_038540.2 |
| AK140485.1 | 1 | 340 | BAE24405.1 | |
| AK161182.1 | 2 | 416 | ||
| AK191189.1 | ||||
| BC007166.1 | 2 | 380 | AAH07166.1 | |
| BC013557.1 | 2 | 380 | AAH13557.1 | |
| CB246498.1 |

Genomic Map