Assay Details
Target Gene Details
Entrez Gene ID: | 13824 |
Gene Name: | erythrocyte protein band 4.1 like 4a |
Gene Aliases: |
Epb4.1l4, Epb41l4a, NBL4 |
Location: |
Chr.18:33796327-34007678 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epb4.1l4a | NM_013512.2 | 24 | 3673 | NP_038540.2 |

Genomic Map