Assay Details
Target Gene Details
Entrez Gene ID: | 13831 |
Gene Name: | enhancer of polycomb homolog 1 (Drosophila) |
Gene Aliases: |
2400007E14Rik, 5730566F07Rik, A930032N02Rik, mir-1893 |
Location: |
Chr.18:6435951-6516108 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epc1 | NM_001276350.1 | 1 | 263 | NP_001263279.1 |
| NM_007935.2 | NP_031961.1 | |||
| NM_027497.3 | 1 | 263 | NP_081773.1 | |
| AF079765.1 | AAC64272.1 | |||
| AK010295.1 | 1 | 224 | ||
| AK017858.1 | 1 | 251 | ||
| AK040254.1 | 1 | 263 | BAC30552.1 | |
| AK148194.1 | BAE28408.1 | |||
| AK160362.1 | 1 | 225 | ||
| BC055805.1 | 1 | 243 | AAH55805.1 | |
| BC059708.1 | 1 | 263 | AAH59708.1 | |
| BC060145.1 | 1 | 243 | AAH60145.1 | |
| BC138622.1 | 1 | 91 | ||
| BC138623.1 | 1 | 91 | ||
| BM936825.1 |
Target Gene Details
Entrez Gene ID: | 100316773 |
Gene Name: | microRNA 1893 |
Gene Aliases: |
Mirn1893, mmu-mir-1893 |
Location: |
Chr.18:6490564-6490646 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir1893 | NR_035446.1 |
More Information
Set Membership: |
|

Genomic Map