Assay Details
Target Gene Details
Entrez Gene ID: | 74644 |
Gene Name: | RIKEN cDNA 4930426D05 gene |
Gene Aliases: |
- |
Location: |
Chr.18:21651565-21656107 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930426D05Rik | NM_001271580.1 | NP_001258509.1 | ||
| AK015205.1 | BAB29745.1 |
Target Gene Details
Entrez Gene ID: | 225266 |
Gene Name: | kelch-like 14 |
Gene Aliases: |
6330403N15Rik, AI785250, printor |
Location: |
Chr.18:21550377-21672932 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klhl14 | NM_001081403.1 | 1 | 641 | NP_001074872.1 |
| AK018108.1 | 1 | 356 | BAB31073.1 | |
| AK173163.1 | 1 | 355 | BAD32441.1 |

Genomic Map