Assay Details
Target Gene Details
Entrez Gene ID: | 107045 |
Gene Name: | leucyl-tRNA synthetase |
Gene Aliases: |
2310045K21Rik, 3110009L02Rik, AW536573, mKIAA1352 |
Location: |
Chr.18:42202349-42262189 on Build GRCm38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lars | NM_134137.2 | 32 | 3503 | NP_598898.2 |
| AK129337.1 | 32 | 3495 | BAC98147.1 | |
| AK166827.1 | 32 | 3491 | BAE39051.1 | |
| AK202429.1 | ||||
| BC006060.1 | 18 | 1909 | AAH06060.1 | |
| BC052715.1 | 32 | 3503 | AAH52715.1 |
More Information
Set Membership: |
|

Genomic Map