Assay Details
Target Gene Details
Entrez Gene ID: | 545260 |
Gene Name: | arylsulfatase i |
Gene Aliases: |
BC020108 |
Location: |
Chr.18:60912240-60919504 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arsi | NM_001038499.1 | 1 | 104 | NP_001033588.1 |
| BC138970.1 | 1 | 150 | ||
| BC141169.1 | 1 | 150 |

Genomic Map