Assay Details
Target Gene Details
Entrez Gene ID: | 67526 |
Gene Name: | autophagy related 12 |
Gene Aliases: |
4931423H11Rik, A330058M13Rik, Apg12l, Atg12l |
Location: |
Chr.18:46732417-46741839 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atg12 | NM_026217.3 | 4 | 1099 | NP_080493.2 |
| AK008698.1 | 4 | 1075 | BAB25839.1 | |
| AK016474.2 | 4 | 1078 | BAB30256.1 | |
| AK088255.1 | 1 | 572 | ||
| AK167027.1 | 4 | 1077 | BAE39200.1 | |
| AK177910.1 | 1 | 68 | ||
| BC070470.1 | 4 | 1069 | AAH70470.1 |

Genomic Map