Assay Details
Target Gene Details
Entrez Gene ID: | 70361 |
Gene Name: | lectin, mannose-binding, 1 |
Gene Aliases: |
2610020P13Rik, AI326273, AU043785, C730041J05, ERGIC53, F5F8D, MCFD1, MR60, P58, gp58 |
Location: |
Chr.18:65980739-66002635 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lman1 | NM_027400.3 | 14 | 2913 | NP_081676.1 |
| AK050376.1 | 1 | 1576 | ||
| AK080250.1 | 1 | 1110 | ||
| BC057165.1 | 14 | 2922 | AAH57165.1 | |
| CN536547.1 | 1 | 401 |
More Information
Set Membership: |
|

Genomic Map