Assay Details
Target Gene Details
Entrez Gene ID: | 22268 |
Gene Name: | uroplakin 1B |
Gene Aliases: |
AI413235, Tspan20, UPIb, Upk1 |
Location: |
Chr.16:38773184-38800345 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Upk1b | NM_178924.4 | 8 | 1611 | NP_849255.2 |
| AK083101.1 | 8 | 1611 | BAC38760.1 | |
| BB427704.2 | 1 | 547 | ||
| BC046604.1 | 8 | 1589 | AAH46604.1 |

Genomic Map