Assay Details
Target Gene Details
Entrez Gene ID: | 68977 |
Gene Name: | hydroxyacylglutathione hydrolase-like |
Gene Aliases: |
1500017E18Rik, 2810014I23Rik, C330022E15Rik |
Location: |
Chr.17:25779843-25785669 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Haghl | NM_001271433.1 | 9 | 1222 | NP_001258362.1 |
| NM_001271434.1 | 9 | 1224 | NP_001258363.1 | |
| NM_001271435.1 | NP_001258364.1 | |||
| NM_001271436.1 | 8 | 1036 | NP_001258365.1 | |
| NM_001271437.1 | 8 | 1031 | NP_001258366.1 | |
| NM_026897.3 | 8 | 1207 | NP_081173.1 | |
| AI844861.1 | 1 | 115 | ||
| AK005274.1 | 8 | 1203 | BAB23924.1 | |
| AK012748.1 | 9 | 1221 | ||
| AK021220.1 | 8 | 1038 | ||
| AK032999.1 | 5 | 1375 | ||
| AK034798.1 | 9 | 1199 | ||
| AK047517.1 | BAC33079.1 | |||
| AK079632.1 | 5 | 1695 | ||
| AK080454.1 | 1 | 2684 | ||
| AK154248.1 | 8 | 1012 | ||
| BC030466.1 | 9 | 1207 | AAH30466.1 | |
| BC083322.1 | 9 | 1161 | AAH83322.1 |
Target Gene Details
Entrez Gene ID: | 67563 |
Gene Name: | nuclear prelamin A recognition factor-like |
Gene Aliases: |
9030612I22Rik, AI504405, PRN |
Location: |
Chr.17:25773759-25783332 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Narfl | NM_026238.4 | 11 | 2108 | NP_080514.3 |
| AK043694.1 | 11 | 2085 | BAC31620.1 |
More Information
Set Membership: |
|

Genomic Map