Assay Details
Target Gene Details
Entrez Gene ID: | 19133 |
Gene Name: | peripherin 2 |
Gene Aliases: |
AOFMD, AVMD, Nmf193, PRPH, RP7, Rd-2, Rd2, Rds, Tspan22, rds |
Location: |
Chr.17:46910485-46924926 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prph2 | NM_008938.1 | 3 | 2106 | NP_032964.1 |
| AK136280.1 | 5 | 2200 | BAE22911.1 | |
| X14770.1 | 3 | 2106 | CAA32878.1 |

Genomic Map