Assay Details
Target Gene Details
Entrez Gene ID: | 260297 |
Gene Name: | proline-rich transmembrane protein 1 |
Gene Aliases: |
Bat-4, Bat4, D17H6S54E, DSPD1, G5b, NG5, ORF31 |
Location: |
Chr.17:34629569-34633134 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prrt1 | NM_030890.1 | 2 | 202 | NP_112152.1 |
| BC115512.2 | ||||
| BC115513.2 |

Genomic Map