Assay Details
Target Gene Details
Entrez Gene ID: | 66306 |
Gene Name: | family with sequence similarity 53, member C |
Gene Aliases: |
2810012G03Rik, Gm21179 |
Location: |
Chr.18:34758830-34773760 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam53c | NM_175104.4 | 5 | 1839 | NP_780313.1 |
| AK044469.1 | 5 | 1643 | BAC31938.1 | |
| AK135712.1 | 4 | 1585 | BAE22622.1 | |
| AK154214.1 | 5 | 1643 | BAE32439.1 | |
| AK166841.1 | 4 | 1583 | ||
| AK213951.1 | 1 | 34 | ||
| BC044896.1 | 4 | 1564 | AAH44896.1 | |
| BC057111.1 | 5 | 1839 | AAH57111.1 |

Genomic Map