Assay Details
Target Gene Details
Entrez Gene ID: | 68092 |
Gene Name: | nuclear cap binding protein subunit 2 |
Gene Aliases: |
20kDa, 5930413E18Rik, AA536802, AI843301, C79367 |
Location: |
Chr.16:31948546-31958472 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncbp2 | NM_026554.4 | 4 | 1773 | NP_080830.1 |
| AI843301.1 | ||||
| AK077841.1 | BAC37030.1 | |||
| AK078181.1 | 4 | 1704 | ||
| AK135597.1 | ||||
| AK147027.1 | 4 | 1705 | BAE27620.1 | |
| AK184640.1 |
More Information
Set Membership: |
|

Genomic Map