Assay Details
Target Gene Details
Entrez Gene ID: | 53858 |
Gene Name: | RWD domain containing 2B |
Gene Aliases: |
ORF5 |
Location: |
Chr.16:87433331-87440592 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rwdd2b | NM_016924.2 | 5 | 1017 | NP_058620.2 |
| AF177771.1 | 5 | 973 | AAF63681.1 | |
| AK002746.1 | 5 | 983 | ||
| AK084966.1 | 5 | 992 | BAC39322.1 | |
| BC002143.1 | 5 | 998 | AAH02143.1 |
More Information
Set Membership: |
|

Genomic Map