Assay Details
Target Gene Details
Entrez Gene ID: | 66968 |
Gene Name: | perilipin 5 |
Gene Aliases: |
2310076L09Rik, AI415325, AW109675, Lsdp5, MLDP, PAT-1 |
Location: |
Chr.17:56111597-56117606 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plin5 | NM_001077348.1 | 8 | 1677 | NP_001070816.1 |
| NM_025874.3 | 9 | 1623 | NP_080150.2 | |
| AK010202.1 | 9 | 1623 | ||
| AK075872.1 | 9 | 1620 | BAC36019.1 | |
| AY919875.1 | 8 | 1636 | AAX12443.1 | |
| BC024138.1 | 9 | 1603 | ||
| DQ473305.1 | 9 | 1603 |
More Information
Set Membership: |
|

Genomic Map