Assay Details
Target Gene Details
Entrez Gene ID: | 57261 |
Gene Name: | bromodomain containing 4 |
Gene Aliases: |
Brd5, HUNK1, MCAP, WI-11513 |
Location: |
Chr.17:32196272-32284722 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Brd4 | NM_001286630.1 | 14 | 2721 | NP_001273559.1 |
| NM_020508.4 | 14 | 2674 | NP_065254.3 | |
| AF273217.1 | 13 | 2589 | AAG02191.1 | |
| AF461395.1 | 14 | 2650 | AAL67833.1 | |
| AK147545.1 | 14 | 2686 | BAE27987.1 |

Genomic Map