Assay Details
Target Gene Details
Entrez Gene ID: | 57261 |
Gene Name: | bromodomain containing 4 |
Gene Aliases: |
Brd5, HUNK1, MCAP, WI-11513 |
Location: |
Chr.17:32196272-32284722 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Brd4 | NM_001286630.1 | 20 | 5473 | NP_001273559.1 |
| NM_020508.4 | 20 | 5426 | NP_065254.3 | |
| AF461395.1 | 20 | 5402 | AAL67833.1 | |
| AK035000.1 | 4 | 1612 | BAC28909.1 | |
| AK147545.1 | 20 | 5438 | BAE27987.1 | |
| AK162710.1 | 4 | 1594 |

Genomic Map