Assay Details
Target Gene Details
Entrez Gene ID: | 18091 |
Gene Name: | NK2 homeobox 5 |
Gene Aliases: |
Csx, Nkx-2.5, Nkx2.5, tinman |
Location: |
Chr.17:26838664-26844994 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkx2-5 | NM_008700.2 | 2 | 1200 | NP_032726.1 |
| AK142146.1 | 2 | 1105 | BAE24947.1 | |
| BC139299.1 | 2 | 1046 | ||
| BC139303.1 | 2 | 1046 | ||
| L20300.1 | ||||
| X75415.1 | 2 | 1265 | CAA53169.1 |
More Information
Set Membership: |
|

Genomic Map