Assay Details
Target Gene Details
Entrez Gene ID: | 50850 |
Gene Name: | spastin |
Gene Aliases: |
Spg4, mKIAA1083 |
Location: |
Chr.17:74337454-74391115 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spast | NM_001162870.1 | 17 | 3713 | NP_001156342.1 |
| NM_016962.2 | 17 | 3710 | NP_058658.2 | |
| AK129282.1 | 17 | 3687 | BAC98092.1 |
More Information
Set Membership: |
|

Genomic Map