Assay Details
Target Gene Details
Entrez Gene ID: | 71020 |
Gene Name: | spermatogenesis associated, serine-rich 1 |
Gene Aliases: |
1700011H05Rik, 4933400B06Rik, Daip2, Ddip, Srsp1 |
Location: |
Chr.17:45445877-45475194 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spats1 | NM_027649.3 | 8 | 1125 | NP_081925.2 |
| AK005865.1 | BAB24285.1 | |||
| AK016586.1 | BAB30325.1 | |||
| AV045867.2 | 1 | 267 | ||
| AY336501.1 | AAQ88273.1 |

Genomic Map