Assay Details
Target Gene Details
Entrez Gene ID: | 76936 |
Gene Name: | heterogeneous nuclear ribonucleoprotein M |
Gene Aliases: |
2610023M21Rik, AA409009, Hnrpm, mKIAA4193 |
Location: |
Chr.17:33646233-33685458 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpm | NM_001109913.1 | 17 | 2343 | NP_001103383.1 |
| NM_029804.3 | 16 | 2460 | NP_084080.1 | |
| AK011521.1 | 16 | 2406 | BAB27675.1 | |
| AK133911.1 | 15 | 2342 | ||
| AK145343.1 | 17 | 2281 | BAE26377.1 | |
| AK150477.1 | 1 | 334 | BAE29593.1 | |
| AK159848.1 | 17 | 2275 | BAE35426.1 | |
| AK163866.1 | 5 | 1292 | BAE37519.1 | |
| AK166670.1 | 16 | 2481 | BAE38931.1 | |
| AK168347.1 | 17 | 2278 | BAE40284.1 | |
| AK169101.1 | 17 | 2277 | ||
| AK171892.1 | 17 | 2278 | BAE42723.1 | |
| AK220299.1 | 17 | 2303 | BAD90224.1 | |
| BC005758.1 | 5 | 1353 | AAH05758.1 | |
| BC065172.1 | 17 | 2273 | AAH65172.1 |
More Information
Set Membership: |
|

Genomic Map