Assay Details
Target Gene Details
Entrez Gene ID: | 18131 |
Gene Name: | notch 3 |
Gene Aliases: |
AW229011, N3, hpbk |
Location: |
Chr.17:32120893-32166852 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Notch3 | NM_008716.2 | 25 | 4693 | NP_032742.1 |
| X74760.1 | 25 | 4693 | CAA52776.1 |

Genomic Map