Assay Details
Target Gene Details
Entrez Gene ID: | 20807 |
Gene Name: | serum response factor |
Gene Aliases: |
AW049942, AW240594 |
Location: |
Chr.17:46546838-46556870 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srf | NM_020493.2 | 7 | 2927 | NP_065239.1 |

Genomic Map