Assay Details
Target Gene Details
Entrez Gene ID: | 12322 |
Gene Name: | calcium/calmodulin-dependent protein kinase II alpha |
Gene Aliases: |
CaMKII, R74975, mKIAA0968 |
Location: |
Chr.18:60925580-60988152 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Camk2a | NM_001286809.1 | 6 | 3104 | NP_001273738.1 |
| NM_009792.3 | 7 | 3137 | NP_033922.1 | |
| NM_177407.4 | 18 | 3825 | NP_803126.1 | |
| AK083245.1 | 18 | 3826 | BAC38829.1 | |
| AK122410.2 | 18 | 3821 | BAC65692.3 |
More Information
Set Membership: |
|

Genomic Map