Assay Details
Target Gene Details
Entrez Gene ID: | 14423 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 1 |
Gene Aliases: |
- |
Location: |
Chr.18:24205344-24286816 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galnt1 | NM_001160404.1 | 13 | 3533 | NP_001153876.1 |
| NM_013814.3 | 13 | 3621 | NP_038842.3 | |
| AK083391.1 | 13 | 3537 | ||
| AK085878.1 | 1 | 924 | ||
| CX123572.1 |
More Information
Set Membership: |
|

Genomic Map