Assay Details
Target Gene Details
Entrez Gene ID: | 15937 |
Gene Name: | immediate early response 3 |
Gene Aliases: |
AI663993, IEX-1, cI-3, gly96 |
Location: |
Chr.17:35821713-35822911 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ier3 | NM_133662.2 | 2 | 362 | NP_598423.1 |
| AK003466.1 | 2 | 388 | ||
| AK051003.1 | 2 | 392 | BAC34493.1 | |
| AK170477.1 | 2 | 392 | BAE41821.1 | |
| BC006950.1 | 2 | 362 | AAH06950.1 | |
| CT010247.1 | 2 | 356 | CAJ18455.1 | |
| CT010271.1 | 2 | 356 | CAJ18479.1 |

Genomic Map