Assay Details
Target Gene Details
Entrez Gene ID: | 381073 |
Gene Name: | neuropeptide W |
Gene Aliases: |
Gm935 |
Location: |
Chr.17:24657330-24658425 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npw | NM_001099664.1 | 1 | 289 | NP_001093134.1 |
| AK131881.1 | 1 | 71 | BAE20848.1 |

Genomic Map