Assay Details
Target Gene Details
Entrez Gene ID: | 18209 |
Gene Name: | netrin 3 |
Gene Aliases: |
Ntn2l |
Location: |
Chr.17:24203831-24209387 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ntn3 | NM_010947.3 | 1 | 1113 | NP_035077.1 |
| AF128866.1 | 1 | 953 | AAD28603.1 | |
| AF149094.1 | 1 | 749 | AAD42078.1 | |
| AF152418.1 | 1 | 868 | AAD40063.1 | |
| AK031199.1 | 1 | 1258 | BAC27297.1 | |
| AK083904.1 | ||||
| AK141856.1 | 1 | 1024 | BAE24860.1 | |
| BC094362.1 | 1 | 1113 | AAH94362.1 |
More Information
Set Membership: |
|

Genomic Map