Assay Details
Target Gene Details
Entrez Gene ID: | 18209 |
Gene Name: | netrin 3 |
Gene Aliases: |
Ntn2l |
Location: |
Chr.17:24203831-24209387 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ntn3 | NM_010947.3 | 6 | 3878 | NP_035077.1 |
| AK031199.1 | 6 | 4023 | BAC27297.1 | |
| AK083904.1 | 6 | 3612 | ||
| AK141856.1 | 6 | 3789 | BAE24860.1 | |
| AK172594.1 | 1 | 759 | ||
| BC094362.1 | 6 | 3878 | AAH94362.1 |
Target Gene Details
Entrez Gene ID: | 224617 |
Gene Name: | TBC1 domain family, member 24 |
Gene Aliases: |
9630033P11, C530046L02Rik, mKIAA1171 |
Location: |
Chr.17:24175431-24205562 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d24 | NM_001163847.1 | NP_001157319.1 | ||
| NM_001163848.1 | NP_001157320.1 | |||
| NM_001163849.1 | NP_001157321.1 | |||
| NM_001163850.1 | NP_001157322.1 | |||
| NM_001163851.1 | NP_001157323.1 | |||
| NM_173186.4 | NP_775278.3 | |||
| AK036088.1 | ||||
| AK045194.1 | BAC32258.1 | |||
| AK049754.1 | BAC33904.1 | |||
| AK083833.1 | BAC39035.1 | |||
| AK084693.1 | BAC39255.1 | |||
| AK086293.1 | BAC39644.1 | |||
| AK129304.1 | BAC98114.1 | |||
| AK138431.1 | BAE23659.1 | |||
| AK147647.1 | BAE28046.1 |
More Information
Set Membership: |
|

Genomic Map