Assay Details
Target Gene Details
Entrez Gene ID: | 19317 |
Gene Name: | quaking |
Gene Aliases: |
1110003F05Rik, 1500005P18, QkI, l(17)-1Wis, l17Wis1 |
Location: |
Chr.17:10202856-10319361 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Qk | NM_001159517.1 | 8 | 2026 | NP_001152989.1 |
| AF090402.1 | 10 | 2117 | AAC99452.1 | |
| AF090403.1 | 10 | 2876 | AAC99453.1 | |
| AK005169.1 | 8 | 1412 | BAB23859.1 | |
| AK010134.1 | 1 | 262 | ||
| AK161210.1 | 1 | 104 | ||
| BC053426.1 | 9 | 1698 | AAH53426.1 | |
| BC056346.1 | 8 | 1552 | AAH56346.1 |
More Information
Set Membership: |
|

Genomic Map