Assay Details
Target Gene Details
Entrez Gene ID: | 19317 |
Gene Name: | quaking |
Gene Aliases: |
1110003F05Rik, 1500005P18, QkI, l(17)-1Wis, l17Wis1 |
Location: |
Chr.17:10202856-10319361 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Qk | NM_001159516.1 | 1 | 523 | NP_001152988.1 |
| NM_001159517.1 | 1 | 523 | NP_001152989.1 | |
| NM_021881.2 | 1 | 523 | NP_068681.1 | |
| AF090402.1 | 2 | 683 | AAC99452.1 | |
| AF090403.1 | 2 | 683 | AAC99453.1 | |
| AK132303.1 | 1 | 524 | BAE21091.1 | |
| BC053426.1 | 2 | 195 | AAH53426.1 | |
| BC056346.1 | 1 | 49 | AAH56346.1 | |
| U44940.1 | 1 | 542 | AAC52491.1 |
More Information
Set Membership: |
|

Genomic Map