Assay Details
Target Gene Details
Entrez Gene ID: | 19317 |
Gene Name: | quaking |
Gene Aliases: |
1110003F05Rik, 1500005P18, QkI, l(17)-1Wis, l17Wis1 |
Location: |
Chr.17:10202856-10319361 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Qk | NM_001159516.1 | 7 | 1906 | NP_001152988.1 |
| NM_001159517.1 | NP_001152989.1 | |||
| NM_021881.2 | 7 | 3186 | NP_068681.1 | |
| AF090402.1 | AAC99452.1 | |||
| AF090403.1 | 8 | 2066 | AAC99453.1 | |
| AK005169.1 | BAB23859.1 | |||
| AK132303.1 | 7 | 1907 | BAE21091.1 | |
| AK142236.1 | 2 | 1797 | BAE24988.1 | |
| AK163757.1 | 1 | 736 | ||
| BC053426.1 | AAH53426.1 | |||
| BC056346.1 | AAH56346.1 | |||
| U44940.1 | 7 | 3205 | AAC52491.1 |
More Information
Set Membership: |
|

Genomic Map