Assay Details
Target Gene Details
Entrez Gene ID: | 106763 |
Gene Name: | tau tubulin kinase 1 |
Gene Aliases: |
AU017937, AW048023, C330008L01Rik |
Location: |
Chr.17:46442448-46487675 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttbk1 | NM_001162864.1 | 15 | 5924 | NP_001156336.1 |
| AK049158.1 | 1 | 886 | BAC33576.1 | |
| AK173279.1 | 2 | 2887 | BAD32557.1 | |
| BC059249.1 | 2 | 3128 |
More Information
Set Membership: |
|

Genomic Map