Assay Details
Target Gene Details
Entrez Gene ID: | 67242 |
Gene Name: | gem (nuclear organelle) associated protein 6 |
Gene Aliases: |
2610019B15Rik, 2810470M17Rik |
Location: |
Chr.17:80207704-80228497 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gemin6 |
Target Gene Details
Entrez Gene ID: | 67737 |
Gene Name: | tetratricopeptide repeat domain 39D |
Gene Aliases: |
4930560E09Rik |
Location: |
Chr.17:80215914-80217936 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc39d | NM_026351.2 | 1 | 675 | NP_080627.2 |
| AK016184.1 | 1 | 258 | BAB30139.3 | |
| AK016869.1 | 1 | 122 | ||
| BC118946.1 | 1 | 675 | ||
| BC118947.1 | 1 | 675 |

Genomic Map