Assay Details
Target Gene Details
Entrez Gene ID: | 21769 |
Gene Name: | zinc finger, AN1-type domain 3 |
Gene Aliases: |
AW539211, TEG-27, Tex27 |
Location: |
Chr.17:30005039-30210020 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfand3 | NM_148926.2 | 5 | 1947 | NP_683728.1 |
| AK082915.1 | 4 | 1943 | ||
| AK206646.1 | ||||
| AK211117.1 | ||||
| BC024483.1 | 4 | 1608 | AAH24483.1 | |
| BC027161.1 | 4 | 1745 | AAH27161.1 | |
| BC034396.1 | 5 | 1867 | AAH34396.1 | |
| BC083124.1 | 5 | 1947 | AAH83124.1 |

Genomic Map