Assay Details
Target Gene Details
Entrez Gene ID: | 677156 |
Gene Name: | cytochrome P450, family 4, subfamily f, polypeptide 37 |
Gene Aliases: |
Gm9705 |
Location: |
Chr.17:32621319-32636184 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp4f37 | NM_001100187.1 | 13 | 1627 | NP_001093657.1 |
| AK132492.1 | 13 | 1627 | BAE21200.1 |
More Information
Set Membership: |
|

Genomic Map