Assay Details
Target Gene Details
Entrez Gene ID: | 53414 |
Gene Name: | bystin-like |
Gene Aliases: |
Bys, Enp1 |
Location: |
Chr.17:47599331-47611492 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bysl | NM_016859.3 | 1 | 196 | NP_058555.3 |
| AK049195.1 | 1 | 189 | BAC33603.1 | |
| AK051704.1 | 1 | 186 | BAC34727.1 | |
| AK083033.1 | 1 | 181 | BAC38736.1 | |
| AK143027.1 | 1 | 197 | BAE25255.1 |

Genomic Map