Assay Details
Target Gene Details
Entrez Gene ID: | 53414 |
Gene Name: | bystin-like |
Gene Aliases: |
Bys, Enp1 |
Location: |
Chr.17:47599331-47611492 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bysl | NM_016859.3 | 7 | 3434 | NP_058555.3 |
| AK143027.1 | 7 | 3435 | BAE25255.1 |
Target Gene Details
Entrez Gene ID: | 12445 |
Gene Name: | cyclin D3 |
Gene Aliases: |
9230106B05Rik, AA682053, AL024085, AW146355, C78795 |
Location: |
Chr.17:47504738-47599689 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnd3 | NM_001081635.1 | NP_001075104.1 | ||
| NM_001081636.1 | NP_001075105.1 | |||
| NM_007632.2 | NP_031658.1 | |||
| AK083384.1 | ||||
| AK159503.1 | BAE35136.1 | |||
| AK161758.1 | BAE36561.1 | |||
| AK168290.1 | BAE40235.1 | |||
| AK183577.1 | ||||
| AK191154.1 | ||||
| AK191156.1 | ||||
| AK191616.1 | ||||
| AK193125.1 | ||||
| AK193325.1 | ||||
| AK195201.1 | ||||
| AK195467.1 | ||||
| AK196525.1 | ||||
| AK196937.1 | ||||
| AK201151.1 | ||||
| AK204876.1 | ||||
| AK206104.1 | ||||
| AK209470.1 | ||||
| AK211904.1 | ||||
| AK212696.1 | ||||
| AK212987.1 | ||||
| AK213832.1 | ||||
| AK214740.1 | ||||
| AK215975.1 | ||||
| AK216696.1 | ||||
| BC004076.1 | AAH04076.1 | |||
| BC005605.1 | AAH05605.1 | |||
| BQ033424.1 |
More Information
Set Membership: |
|

Genomic Map