Assay Details
Target Gene Details
Entrez Gene ID: | 57440 |
Gene Name: | EH-domain containing 3 |
Gene Aliases: |
Ehd2 |
Location: |
Chr.17:73804841-73832093 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ehd3 | NM_020578.3 | 1 | 202 | NP_065603.2 |
| AF155883.1 | 1 | 173 | AAF19020.1 | |
| AF506002.1 | AAM28633.1 | |||
| AK051717.1 | 1 | 93 | ||
| AK128964.1 | 1 | 181 | ||
| AK141265.1 | 1 | 204 | BAE24623.1 |
More Information
Set Membership: |
|

Genomic Map